Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL5 Monoclonal Antibody (03), Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | CXCL5 |
---|---|
Clone | 03 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543007
![]() |
Thermo Scientific
MA543007 |
100 μL |
Each for $387.00
|
N/A |
PIMA543387
![]() |
Thermo Scientific
MA543387 |
50 μL |
Each for $221.00
|
N/A |
Description
Specificity: Human CXCL5
CXCL5 Monoclonal antibody specifically detects CXCL5 in Human samples. It is validated for ELISASpecifications
CXCL5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
AMCF-II; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5 | |
CXCL5 | |
IgG1 | |
Protein A | |
Antibody |
03 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P42830 | |
6374 | |
Recombinant Human CXCL5 Protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
CXCL5 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title