Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL5 Rat anti-Mouse, Biotin, Clone: 32B3, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222026B
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
CXCL5 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CXCL5 | |
Mouse recombinant CXCL5/ENA-78 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
32B3 | |
Western Blot, ELISA | |
chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
Rat | |
Protein G purified | |
Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
6374 | |
Store at 4C in the dark. | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title