Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL6/GCP-2 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0217-10L25, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212232AF594

Catalog No. NP212232A94


Add to cart

Description

Description

CXCL6/GCP-2 Monoclonal antibody specifically detects CXCL6/GCP-2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CXCL6/GCP-2
MM0217-10L25
Western Blot
Conjugated, Purified
CXCL6
Human GCP2
Protein G purified
Cell Cycle and Replication, Chemokines and Cytokines
Primary
6372.0
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit