Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL7/NAP-2 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0211-23B23, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212234AF532

Catalog No. NP212234A32


Add to cart

Description

Description

CXCL7/NAP-2 Monoclonal antibody specifically detects CXCL7/NAP-2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CXCL7/NAP-2
RM0211-23B23
Western Blot
Conjugated, Purified
PPBP
Mouse NAP2
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines, Immunology, mTOR Pathway
Primary
5473.0
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Beta-TG, connective tissue-activating peptide III, CTAP3CTAP-III, CTAPIII, CXC chemokine ligand 7, C-X-C motif chemokine 7, CXCL7b-TG1, LA-PF4, LDGFTC1, Leukocyte-derived growth factor, low-affinity platelet factor IV, Macrophage-derived growth factor, MDGFTC2, NAP-2, NAP-2-L1, neutrophil-activating peptide 2, neutrophil-activating peptide-2, PBPTHBGB, platelet basic protein, pro-platelet basic protein (chemokine (C-X-C motif) ligand 7), SCAR10, SCYB7beta-thromboglobulin, small inducible cytokine B7, small inducible cytokine subfamily B, member 7, Small-inducible cytokine B7, TGB, TGB1B-TG1, THBGB1, thrombocidin 1, thrombocidin 2, thromboglobulin, beta-1
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit