Learn More
CXCL7/NAP-2 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0211-23B23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212234AF594
Catalog No. NP212234A94
Add to cart
Description
Description
CXCL7/NAP-2 Monoclonal antibody specifically detects CXCL7/NAP-2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CXCL7/NAP-2
RM0211-23B23
Western Blot
Beta-TG, connective tissue-activating peptide III, CTAP3CTAP-III, CTAPIII, CXC chemokine ligand 7, C-X-C motif chemokine 7, CXCL7b-TG1, LA-PF4, LDGFTC1, Leukocyte-derived growth factor, low-affinity platelet factor IV, Macrophage-derived growth factor, MDGFTC2, NAP-2, NAP-2-L1, neutrophil-activating peptide 2, neutrophil-activating peptide-2, PBPTHBGB, platelet basic protein, pro-platelet basic protein (chemokine (C-X-C motif) ligand 7), SCAR10, SCYB7beta-thromboglobulin, small inducible cytokine B7, small inducible cytokine subfamily B, member 7, Small-inducible cytokine B7, TGB, TGB1B-TG1, THBGB1, thrombocidin 1, thrombocidin 2, thromboglobulin, beta-1
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
PPBP
Mouse NAP2
0.1 mL
Primary
5473