Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL8/IL-8 Mouse anti-Human, Clone: 1028336, R&D Systems™

Mouse Monoclonal Antibody

$122.30 - $314.00

Specifications

Antigen CXCL8/IL-8
Clone 1028336
Dilution Western Blot 3 ug/mL, Simple Western 50 ug/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2083100
View Documents
R&D Systems
MAB2083100
100 μg
Each for $314.00
Add to cart
 
MAB2083SP
View Documents
R&D Systems
MAB2083SP
25 μg
Each for $122.30
Add to cart
 
Description

Description

CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Western Blot
Specifications

Specifications

CXCL8/IL-8
Western Blot 3 ug/mL, Simple Western 50 ug/mL
Unconjugated
Mouse
Human
P10145.1
3576
IgG1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1028336
Western Blot
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
E. coli-derived human IL-8/CXCL8, Ser28-Ser99, Accession # P10145.1
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit