Learn More
Abnova™ CXCL9 (23-126) 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00004283T04
Catalog No. 89-941-088
Add to cart
Specifications
Specifications
Western Blot
293T
BC063122
pCMV-CXCL9 (23-126) partial fragment
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CXCL9 (23-126) 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 11.44kDa
Transient overexpression cell lysate was tested with anti-CXCL9 (23-126) antibody (Mfr. No. H00004283-B02P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)