Learn More
CXCL9/MIG Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0220-7F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212236AF532
Catalog No. NP212236A32
Add to cart
Description
Description
CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
CXCL9/MIG
MM0220-7F11
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CXCL9
Human MIG
0.1 mL
Primary
4283