Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL9/MIG Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0220-7F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212236AF750

Catalog No. NP212236A70


Add to cart

Description

Description

CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

CXCL9/MIG
MM0220-7F11
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
CXCL9
Human MIG
Protein G purified
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Primary
4283.0
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit