Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9/MIG Rabbit anti-Human, Clone: 50, Novus Biologicals™
Rabbit Monoclonal Antibody
Specifications
Antigen | CXCL9/MIG |
---|---|
Clone | 50 |
Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
CXCL9/MIG | |
ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
Recombinant Human CXCL9/MIG protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
50 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
0.2 um filtered solution in PBS | |
4283 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title