Learn More
CXCR1/IL-8RA Mouse anti-Human, Hamster, PerCP, Clone: 8F1-1-4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143339PCP
Catalog No. NBP143339JP
Add to cart
Description
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Human, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications
Specifications
CXCR1/IL-8RA
8F1-1-4
Flow Cytometry, Immunohistochemistry-Paraffin
Purified
CXCR1
The immunogen for this antibody was CD181.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin)
PerCP
PBS with 0.05% Sodium Azide
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Mouse
IgG2b Kappa
0.1 ml
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
3577