Learn More
CXCR1/IL-8RA Mouse anti-Human, Alexa Fluor 647, Clone: 42709, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3302R100UG
Catalog No. FAB3302R100
Add to cart
This item is not returnable. View return policy
Description
Description
CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCR1/IL-8RA
42709
Flow Cytometry 0.25-1 ug/10^6 cells
AAA59159
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA, Met1-Leu350, Accession # AAA59159
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3577
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG2a
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human