Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR2/IL-8RB Mouse anti-Human, Biotin, Clone: 5E8-C7-F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143338B

Catalog No. NBP143338B


Add to cart

Description

Description

CXCR2/IL-8RB Monoclonal antibody specifically detects CXCR2/IL-8RB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

CXCR2/IL-8RB
5E8-C7-F10
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CXCR2
The immunogen for this antibody was CD182.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Mouse
IgG1 Kappa
0.1 ml
Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary
3579
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit