Learn More
CXCR2/IL-8RB Antibody (242216), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2164MFV500
Description
CXCR2/IL-8RB Monoclonal antibody specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-reportedSpecifications
| CXCR2/IL-8RB | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| 3579 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| 242216 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-reported | |
| CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
| C6 rat glioma cell line transfected with mouse CXCR2, Met1-Leu359, Accession # P35343 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |