Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR2/IL-8RB Rat anti-Mouse, Clone: RM0212-24D12, Novus Biologicals™
Rat Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | CXCR2/IL-8RB |
---|---|
Clone | RM0212-24D12 |
Concentration | LYOPH |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Blocking Assay, CyTOF |
Classification | Monoclonal |
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready, Block/Neutralize.Specifications
CXCR2/IL-8RB | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
CXCR2 | |
Primary |
RM0212-24D12 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Blocking Assay, CyTOF | |
Unconjugated | |
Rat | |
Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
P35343 | |
3579 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title