Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 Rat anti-Mouse, PE/Cy5.5, Clone: RM0213-14C23, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP212239PECY55
Description
CXCR3 Monoclonal antibody specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry.Specifications
CXCR3 | |
RM0213-14C23 | |
Flow Cytometry | |
Conjugated, Purified | |
CXCR3 | |
Mouse CXCR3 | |
Protein G purified | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
Primary | |
2833.0 |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
Rat | |
IgG2 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title