Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112543
Description
In Western blot, this antibody detects a band at ∽42 kDa. A suggested positive control for this product is HeLa cell lysate.
Human immunodeficiency virus (HIV) and related viruses require coreceptors, in addition to T-cell antigen T4 (CD4), to infect target cells. Some G protein-coupled receptors including chemokine (C-C) receptor 5 (CCR5), chemokine (C-X-C) receptor 4 (CXCR4), CCR3, CCR2b, and CCR8 in the chemokine receptor family, and four new human molecules G-protein coupled receptor 15 (GPR15), CXCR6, GPR1, and chemokine (C-X3-C) receptor 1 (CX3CR1) were recently identified as HIV coreceptors. Among them, CXCR4 (fusin, LESTR, or HUMSTR) is a principal coreceptor for T-cell tropic strains of HIV-1 fusion and entry of human white blood cells. CXCR4 is also required for the infection by dual-tropic strains of HIV-1 and mediates CD-4 independent infection by HIV-2. The alpha-chemokine stromal cell-derived factor-1 (SDF-1) is the ligand for CXCR4 and prevents infection by T-tropic HIV-1. CXCR4 associates with the surface CD4-gp120 complex before HIV enters target cells. CXCR4 messenger RNA levels correlate with HIV-1 permissiveness in diverse human cell types. Some antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV binding sites.
Specifications
CXCR4 | |
Polyclonal | |
Unconjugated | |
CXCR4 | |
D2S201E, FB22, HM89, HSY3RR, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, C-X-C chemokine receptor type 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12767, 7852 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P61073, P70658 | |
CXCR4 | |
Synthetic peptide corresponding to residues 1-14 of the N terminal of human CXCR4 receptor. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction