Learn More
CXCR4 Rat anti-Mouse, APC, Clone: 247506, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB21651A100UG
Catalog No. FB21651A100
Add to cart
This item is not returnable. View return policy
Description
Description
CXCR4 Monoclonal antibody specifically detects CXCR4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCR4
247506
Flow Cytometry 0.25-1 μg/106 cells
P70658
CXCR4
Y3 rat myeloid cell line transfected with mouse CXCR4, Met1-Ser359, Accession No. P70658
100 μg
Primary
7852
Mouse
Flow Cytometry
APC
Supplied in a saline solution containing BSA and sodium azide
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS
Rat
IgG2b
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse CXCR4 transfectants. Does not stain irrelevant transfectants.