Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR5 Mouse anti-Human, PE-Cyanine7, Clone: MM0225-8C17, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212242PECY7

Catalog No. N212242PEC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CXCR5 Monoclonal antibody specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CXCR5
MM0225-8C17
Flow Cytometry
Conjugated, Purified
CXCR5
Human CXCR5
Protein G purified
Adaptive Immunity, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary
643.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Mouse
IgG2
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit