Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR6 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0226-2B44, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212243AF350

Catalog No. NP212243A30


Add to cart

Description

Description

CXCR6 Monoclonal antibody specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

CXCR6
MM0226-2B44
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
CXCR6
Human CXCR6
Protein G purified
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary
10663.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Mouse
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit