Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR6 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB699R100UG
This item is not returnable.
View return policy
Description
CXCR6 Monoclonal specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O00574 | |
CXCR6 | |
NS0 mouse myeloma cell line transfected with human CXCR6 Met1-Leu342 Accession # O00574 | |
RUO | |
10663 | |
Human | |
IgG2b |
Flow Cytometry | |
56811 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
Mouse | |
100 μg | |
Primary | |
Stains human CXCR6 transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction