Learn More
CXCR6 Mouse anti-Rat, Alexa Fluor 750, Clone: 879112, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8196S100UG
Catalog No. FAB8196S100
Add to cart
This item is not returnable. View return policy
Description
Description
CXCR6 Monoclonal antibody specifically detects CXCR6 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCR6
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Mouse
IgG1
RUO
Primary
10663
Rat
Flow Cytometry
879112
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CXCR6
NS0 mouse myeloma cell line transfected with rat CXCR6
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects rat CXCR6 in ELISA. Stains HEK293 cells transfected with rat CXCR6 by Flow Cytometry, but does not stain irrelevant transfectants.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)