Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR6 Rat anti-Mouse, PE/Atto594, Clone: RM0214-16A4, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP212244PEATT594
Description
CXCR6 Monoclonal antibody specifically detects CXCR6 in Mouse samples. It is validated for Flow Cytometry.Specifications
CXCR6 | |
RM0214-16A4 | |
Flow Cytometry | |
Conjugated, Purified | |
CXCR6 | |
Mouse CXCR6 | |
Protein G purified | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
Primary | |
10663.0 |
Flow Cytometry | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
Rat | |
IgG2 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title