Learn More
CXCR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148401AP
Catalog No. 50-172-9768
Add to cart
Description
Description
This antibody is specific to CXCR7.
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
Specifications
Specifications
CXCR7
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O89039, P25106, P56485
Chemokine orphan receptor 1, CMKOR1, CXC R7, CXCR 7, CXCR7, G protein coupled receptor 159, GPR159, RDC 1, RDC1
Rabbit
IgG
150 μL
-20°C
Primary
12778, 57007, 84348
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ACKR3
Liquid
ACKR3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat