Learn More
CXCR7/RDC-1 Mouse anti-Rat, Alexa Fluor 594, Clone: 896032, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8399T100UG
Catalog No. FAB8399T100
Add to cart
This item is not returnable. View return policy
Description
Description
CXCR7/RDC-1 Monoclonal antibody specifically detects CXCR7/RDC-1 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCR7/RDC-1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_445804
CXCR7
NS0 mouse myeloma cell line transfected with rat CXCR7/RDC-1 Met1-Lys362 Accession # NP_445804
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains rat CXCR7/RDC-1 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
896032
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Mouse
IgG2a
RUO
Primary
57007
Rat
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)