Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR7/RDC-1 Rabbit anti-Human, Mouse, Rat, Sheep, PE/Atto594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224779PEATT594

Catalog No. N224779P594


Add to cart

Description

Description

CXCR7/RDC-1 Polyclonal antibody specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Flow Cytometry.
Specifications

Specifications

CXCR7/RDC-1
PE/ATTO 594
PBS with 0.05% Sodium Azide
chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Polyclonal
Human, Mouse, Rat, Sheep
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
ACKR3
A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1.
Peptide affinity purified
Chemokines and Cytokines, GPCR
Primary
57007.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit