Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXorf26 Mouse anti-Human, Clone: OTI4E11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA503132
Description
PBDC1 is a protein coding gene.Specifications
CXorf26 | |
Monoclonal | |
0.48 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BVG4 | |
PBDC1 | |
Full length human recombit protein of human CXorf26 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4E11 | |
Unconjugated | |
PBDC1 | |
CXorf26 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51260 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction