Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXorf40A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570744
Description
This target displays homology in the following species: Dog: 93%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%.
CXorf40A may have an important role of cell protection in inflammation reaction.
Specifications
CXorf40A | |
Polyclonal | |
Unconjugated | |
CXorf40A | |
chromosome X open reading frame 40A; CXorf40; CXorf40A; Endothelial-overexpressed lipopolysaccharide-associated factor 1; EOLA1; FLJ16423; Protein CXorf40A | |
Rabbit | |
Affinity chromatography | |
RUO | |
91966 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TE69 | |
CXorf40A | |
synthetic peptide directed towards the C-terminal region of human CXorf40A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction