Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf41 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549290

Catalog No. PA549290

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Plays a role in cytoplasmic pre-assembly of axonemal dynein.
Specifications

Specifications

CXorf41
Polyclonal
PBS with 0.09% sodium azide
Q9NQM4
Protein PIH1D3, PIH1 domain-containing protein 3, Sarcoma antigen NY-SAR-97, PIH1D3, CXorf41
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
139212
Western Blot
Unconjugated
PIH1D3
Liquid
PIH1D3
KLH conjugated synthetic peptide between 171-199 amino acids from the C-terminal region of human CXorf41.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit