Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf66 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP17051120UL

 View more versions of this product

Catalog No. NBP170520UL


Add to Cart

Description

Description

CXorf66 Polyclonal specifically detects CXorf66 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CXorf66
Polyclonal
Western Blot 1:100-1:2000
chromosome X open reading frame 66, hypothetical protein LOC347487
Rabbit
40 kDa
20 μL
Primary
Human
IgG
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
CXORF66
Synthetic peptides corresponding to CXorf66(chromosome X open reading frame 66) The peptide sequence was selected from the middle region of CXorf66. Peptide sequence PLYSSHPQNEISPSKPFGPQELAKPPKHFNPKRSVSLGRAALLSNSELAE.
Affinity Purified
RUO
347487
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit