Learn More
GE Healthcare CY2 BIS NHS ESTER
Used for high-throughput screening applications
Manufacturer:  GE Healthcare PA12004
Catalog No. 45-999-000
Add to cart
Specifications
Specifications
Red
2 - 8°C, protected from light
949.11 
Used for high-throughput screening (HTS) applications
10 mg