Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Cy3 NHS Ester Mono-Reactive CyDye Available on GSA/VA Contract for Federal Government customers only.

Use for highly accurate labeling of amine groups. GE Healthcare Cy3 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

Manufacturer:  GE Healthcare PA13102

 View more versions of this product

Catalog No. 45-100-200


Add to cart

Description

Description

CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.

Cy3 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Red
For labelling via amine groups. CyDyes are suitable for use in applications such as fluorescence resonance energy transfer (FRET), time resolved- fluorescence resonance energy transfer (TR-FRET), and fluorescence polarization (FP)
Foil packs of pre-measured dried dye
Cy3 NHS Ester
50 mg
570 nm
Dried
Cy3
3 to 10
2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
765.95
Soluble in cold water and hot water
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit