Learn More
Aat BioquestSupplier Diversity Partner Cy5 tertrazine [Cy5 tertrazine] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 911
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.
Catalog No. 50-195-2959