Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanamide, 95%, stabilized, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
420-04-2
CH2N2
42.04
MFCD00007572
XZMCDFZZKTWFGF-UHFFFAOYSA-N
hydrogen,carbimide,amidocyanogen,carbamonitrile,cyanoamine,cyanogenamide,cyanogen nitride,n-cyanoamine,alzogur,dormex
9864
CHEBI:16698
cyanamide
C(#N)N
420-04-2
CH2N2
42.04
MFCD00007572
XZMCDFZZKTWFGF-UHFFFAOYSA-N
hydrogen,carbimide,amidocyanogen,carbamonitrile,cyanoamine,cyanogenamide,cyanogen nitride,n-cyanoamine,alzogur,dormex
9864
CHEBI:16698
cyanamide
C(#N)N
Color White
Flash Point 141°C
Packaging Glass bottle
Infrared Spectrum Authentic
Stabilizer 0.1% (typically) formic acid
Quantity 25g
Melting Point 40.0°C to 47.0°C
Boiling Point 260.0°C
Assay Percent Range 94% min. (Argentometry)
Linear Formula H2NCN
Beilstein 03, IV, 145
Fieser 01,170; 03,71; 05,168; 12,147
Merck Index 15, 2674
Solubility Information Solubility in water: 775g/L. Other solubilities: soluble in alchol,phenols,amines,ethers,ketones,slightly soluble in benzene and halogenated,hydrocarbons
Formula Weight 42.04
Physical Form Crystals
Percent Purity 95%
Chemical Name or Material Cyanamide, 95%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Suspecte

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 206-992-3

Recommended Storage : Refrigerator +4°C