Cyanide, Standard solution, Specpure, CN 1000µg/mL, Alfa Aesar™

Manufacturer: Alfa Aesar 43804AP

Quantity 500mL

 View more versions of this product

Catalog No. AA43804AP

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula KCN in 0.1% NaOH
    MDL Number MFCD00468042
    Chemical Name or Material Cyanide, Standard Solution

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 500mL
    Molecular Formula KCN in 0.1% NaOH
    MDL Number MFCD00468042
    Concentration or Composition (by Analyte or Components) CN: 1000μg/mL
    Health Hazard 1 H301-H311-H332: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled.
    Health Hazard 2 Keep container in a well ventilated place. Wear suitable protective clothing and gloves.
    Odor Odorless
    Chemical Name or Material Cyanide, Standard Solution

    Keep container in a well ventilated place. Wear suitable protective clothing and gloves.

    H301-H311-H332: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit