Learn More
Aat BioquestSupplier Diversity Partner Cyanine 3.5 amine [equivalent to Cy3.5® amine] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 139
A variety of cyanine 3.5 (Cy3.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. They are widely used for labeling peptides, proteins and oligos etc.
Catalog No. 50-195-2749