Learn More
Aat BioquestSupplier Diversity Partner Cyanine 3 bissuccinimidyl ester, potassium salt [same as GE Cy3® bisNHS ester] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 270
This Cy3® dye is the same molecule to GE's Cy3® bisNHS ester. It has two NHS groups that might give brighter signal on some proteins than the Cy3 mono NHS ester (#141 and #271).
Catalog No. 50-195-2824