Learn More
Aat BioquestSupplier Diversity Partner Cyanine 5.5 bissuccinimidyl ester, potassium salt [same as GE Cy5.5® bisNHS ester] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 285
This Cy5.5® dye is the same molecule to GE's Cy5.5® bisNHS ester. It readily reacts with amino groups. It has two NHS groups that might give brighter signal on some proteins than the Cy5.5 mono NHS ester (#174 and #283).
Catalog No. 50-195-2830