Learn More
Aat BioquestSupplier Diversity Partner Cyanine 7 hydrazide [equivalent to Cy7® hydrazide] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 166
A variety of cyanine 7 (Cy7®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. They are widely used for labeling peptides, proteins and oligos etc.
Catalog No. 50-195-2779