Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Cyano-4-hydroxycinnamic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
28166-41-8
C10H7NO3
189.17
MFCD00004204
AFVLVVWMAFSXCK-YVMONPNESA-N
alpha-cyano-4-hydroxycinnamate,2-cyano-4-hydroxycinnamate,2-cyano-3-4-hydroxyphe-nyl acrylic acid,2z-2-cyano-3-4-hydroxyphenyl prop-2-enoic acid
9794103
(Z)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=C(C#N)C(=O)O)O
28166-41-8
C10H7NO3
189.17
MFCD00004204
AFVLVVWMAFSXCK-YVMONPNESA-N
alpha-cyano-4-hydroxycinnamate,2-cyano-4-hydroxycinnamate,2-cyano-3-4-hydroxyphe-nyl acrylic acid,2z-2-cyano-3-4-hydroxyphenyl prop-2-enoic acid
9794103
(Z)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=C(C#N)C(=O)O)O
Color Yellow
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC6H4CH=C(CN)CO2H
Beilstein 10, 520
Formula Weight 189.17
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material α-Cyano-4-hydroxycinnamic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning