Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Cyano-o-tolunitrile (Tech.), ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3759-28-2
C9H6N2
142.16
MFCD00001895
GKHSEDFDYXZGCG-UHFFFAOYSA-N
2-cyanomethyl benzonitrile,homophthalonitrile,2-cyanobenzyl cyanide,2-cyanophenylacetonitrile,alpha-cyano-o-tolunitrile,o-cyanobenzyl cyanide,benzeneacetonitrile, 2-cyano,2-cyanobenzeneacetonitrile,o-cyanophenyl acetonitrile,o-tolunitrile, alpha-cyano
77368
2-(cyanomethyl)benzonitrile
C1=CC=C(C(=C1)CC#N)C#N
3759-28-2
C9H6N2
142.16
MFCD00001895
GKHSEDFDYXZGCG-UHFFFAOYSA-N
2-cyanomethyl benzonitrile,homophthalonitrile,2-cyanobenzyl cyanide,2-cyanophenylacetonitrile,alpha-cyano-o-tolunitrile,o-cyanobenzyl cyanide,benzeneacetonitrile, 2-cyano,2-cyanobenzeneacetonitrile,o-cyanophenyl acetonitrile,o-tolunitrile, alpha-cyano
77368
2-(cyanomethyl)benzonitrile
C1=CC=C(C(=C1)CC#N)C#N
Melting Point 76°C
Packaging Glass bottle
Quantity 100g
Color Beige to Brown
Infrared Spectrum Authentic
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Linear Formula NCCH2C6H4CN
Beilstein 09, 859
Formula Weight 142.16
Physical Form Powder and/or Chunks
Percent Purity ≥75%
Grade Technical
Chemical Name or Material α-Cyano-o-tolunitrile, tech.
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 223-167-3

RTECSNumber : XV0930000

TSCA : TSCA