Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanocobalamin, Crystalline, USP, 96-102%, Spectrum™

Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
68-19-9
C63H89CoN14O14P
1356.40
AGVAZMGAQJOSFJ-UHFFFAOYNA-M
λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-[2-({2-[({[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}(hydroxy)phosphoryl)oxy]propyl}carbamoyl)ethyl]-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide iminomethanide
[Co++].[C-]#N.CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP(O)(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12
68-19-9
C63H89CoN14O14P
1356.40
AGVAZMGAQJOSFJ-UHFFFAOYNA-M
λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-[2-({2-[({[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}(hydroxy)phosphoryl)oxy]propyl}carbamoyl)ethyl]-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide iminomethanide
[Co++].[C-]#N.CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP(O)(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12

CY105, 68-19-9, C63H88CoN14O14P

Spectrum™ Cyanocobalamin, Crystalline, USP is a man-made form of vitamin B12 used to treat low levels (deficiency) of this vitamin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 12%
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 100%
Percent Purity 96 to 102%