Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen bromide, 3M solution in dichloromethane, ACROS Organics™

Click to view available options
Quantity:
100mL
506-68-3,75-09-2
CBrN
105.922
MFCD00011597
ATDGTVJJHBUTRL-UHFFFAOYSA-N
cyanogen bromide,bromine cyanide,bromocyanide,cyanobromide,bromocyanogen,bromocyan,campilit,cyanogen monobromide,bromure de cyanogen,cyanogen bromide brcn
10476
carbononitridic bromide
C(#N)Br
506-68-3,75-09-2
CBrN
105.922
MFCD00011597
ATDGTVJJHBUTRL-UHFFFAOYSA-N
cyanogen bromide,bromine cyanide,bromocyanide,cyanobromide,bromocyanogen,bromocyan,campilit,cyanogen monobromide,bromure de cyanogen,cyanogen bromide brcn
10476
carbononitridic bromide
C(#N)Br
C-X bond formation reagent
Concentration 2.95 to 3.30M
Fieser 01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
Density 1.4400g/mL
Merck Index 15, 2683
Formula Weight 105.93
Physical Form Solution
Specific Gravity 1.44
Color Colorless to Yellow
Quantity 100mL
Chemical Name or Material Cyanogen bromide, 3M Solution in Dichloromethane

GHS H Statement
Fatal if swallowed. Causes severe skin burns and eye damage. Fatal if inhaled. Fatal in contact with skin. Suspected of causing cancer. Very toxic to aquatic life with long lasting effects. May cause drowsiness or dizziness.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF exposed or concerned: Get medical advice/attention.

Danger