Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass Bottle in Tin Can
Glass Bottle packed in tin can
506-68-3
CBrN
105.922
MFCD00011597
ATDGTVJJHBUTRL-UHFFFAOYSA-N
cyanogen bromide,bromine cyanide,bromocyanide,cyanobromide,bromocyanogen,bromocyan,campilit,cyanogen monobromide,bromure de cyanogen,cyanogen bromide brcn
10476
carbononitridic bromide
C(#N)Br
506-68-3
CBrN
105.922
MFCD00011597
ATDGTVJJHBUTRL-UHFFFAOYSA-N
cyanogen bromide,bromine cyanide,bromocyanide,cyanobromide,bromocyanogen,bromocyan,campilit,cyanogen monobromide,bromure de cyanogen,cyanogen bromide brcn
10476
carbononitridic bromide
C(#N)Br
C-X bond formation reagent
Quantity 25g
Density 1.44
Packaging Glass Bottle in Tin Can
Color White
Assay Percent Range 97%
Linear Formula BrCN
Fieser 01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
Merck Index 15, 2683
Formula Weight 105.93
Physical Form Crystals
Percent Purity ≥96.0%
Chemical Name or Material Cyanogen bromide, 97%
Show More Show Less

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Refrigerator +4°C