Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanuric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
Packaging:
Plastic bottle
108-80-5
C3H3N3O3
129.075
MFCD00082990
ZFSLODLOARCGLH-UHFFFAOYSA-N
cyanuric acid,isocyanuric acid,1,3,5-triazine-2,4,6-triol,trihydroxycyanidine,tricyanic acid,sym-triazinetriol,s-triazinetriol,1,3,5-triazine-2,4,6 1h,3h,5h-trione,pseudocyanuric acid,isocyanurate acid
7956
CHEBI:38028
1,3,5-triazinane-2,4,6-trione
C1(=O)NC(=O)NC(=O)N1
108-80-5
C3H3N3O3
129.075
MFCD00082990
ZFSLODLOARCGLH-UHFFFAOYSA-N
cyanuric acid,isocyanuric acid,1,3,5-triazine-2,4,6-triol,trihydroxycyanidine,tricyanic acid,sym-triazinetriol,s-triazinetriol,1,3,5-triazine-2,4,6 1h,3h,5h-trione,pseudocyanuric acid,isocyanurate acid
7956
CHEBI:38028
1,3,5-triazinane-2,4,6-trione
C1(=O)NC(=O)NC(=O)N1
Assay Percent Range 98%
Beilstein 26, 239
Merck Index 15, 2690
Formula Weight 129.07
Physical Form Crystalline Powder or Lumps
Percent Purity ≥97.5%
Infrared Spectrum Authentic
Packaging Plastic bottle
Color White to Beige
Quantity 25g
Chemical Name or Material Cyanuric acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 203-618-0

RTECSNumber : XZ1800000

TSCA : TSCA