Learn More
Aviva Systems Biology Corp CYB5D1 Antibody - middle region (ARP52699_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYB5D1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYB5D1. Alternate names: FLJ32499ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-60808