Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5D2 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548159

Catalog No. PA548159

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and chicken based on sequence homology.

Heme-binding protein which promotes neuronal but not astrocyte differentiation.
Specifications

Specifications

CYB5D2
Polyclonal
PBS with 0.09% sodium azide
Q5SSH8
Neuferricin, Cytochrome b5 domain-containing protein 2, CYB5D2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
192986
Western Blot
Unconjugated
Cyb5d2
Liquid
CYB5D2
KLH conjugated synthetic peptide between 187-216 amino acids from the C-terminal region of human CYB5D2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit