Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R1 Mouse anti-Human, Clone: OTI4B7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504564

Catalog No. 50-167-5222


Add to cart

Description

Description

NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
Specifications

Specifications

CYB5R1
Monoclonal
0.74 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UHQ9
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51706
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4B7
Unconjugated
CYB5R1
Liquid
CYB5R1
Full length human recombit protein of human CYB5R1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit