Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB5R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553128
Description
Immunogen sequence: GGTGITPMLQ LIRAILKVPE DPTQCFLLFA NQTEKDIILR EDLEELQARY PNRFKLWFTL DHPPKDWAYS KGFVTADMIR EHLPAPGDDV LVLLCGPPPM VQLACHPNLD KLGYSQKMRF TY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
Specifications
CYB5R1 | |
Polyclonal | |
Unconjugated | |
CYB5R1 | |
1500005G05Rik; b5R.1; B5R1; B5R2; C80155; Cyb5r1; cytochrome b5 reductase 1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; cytochrome b5 reductase 1; NADH-cytochrome b5 reductase 1; Nqo3a2; UNQ3049/PRO9865 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51706 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHQ9 | |
CYB5R1 | |
Recombinant protein corresponding to Human CYB5R1. Recombinant protein control fragment (Product #RP-90355). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction