Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R2 Mouse anti-Human, APC, Clone: OTI4B9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271942APC
Description
CYB5R2 Monoclonal specifically detects CYB5R2 in Human samples. It is validated for Flow Cytometry.Specifications
CYB5R2 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
CYB5R2 | |
Full length recombinant protein of human CYB5R2 (NP_057313) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry | |
OTI4B9 | |
Flow Cytometry | |
B5R.2, cytochrome b5 reductase 2 | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology | |
51700.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title